Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 16α-hydroxyestrone | 566-76-7 | MFCD00870436 | 99.7% | 286.37 | C18H22O3 | 5 MG
SDP

Catalog No. 5000263527
Encompass_Preferred

16α-hydroxyestrone is an endogenous steroid metabolite of estrone supplied as a solid reference standard for analytical and biochemical research. It is intended for use in metabolism studies, biomarker research, and as a well-characterized compound for assay development.

  • High purity (99.7%).
  • Solid, white to off-white appearance.
  • Molecular formula C18H22O3; molecular weight 286.37.
  • Soluble in DMSO (≥250 mg/mL).
  • Powder storage: -20°C, stable up to 3 years.
  • In solvent: -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-002-63527 Supplier Medchemexpress LLC Supplier No. HY1133445MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

16α-hydroxyestrone is an endogenous steroid metabolite of estrone supplied as a solid reference standard for analytical and biochemical research. It is intended for use in metabolism studies, biomarker research, and as a well-characterized compound for assay development.

  • High purity (99.7%).
  • Solid, white to off-white appearance.
  • Molecular formula C18H22O3; molecular weight 286.37.
  • Soluble in DMSO (≥250 mg/mL).
  • Powder storage: -20°C, stable up to 3 years.
  • In solvent: -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.