Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 16α-hydroxyprednisolone | 13951-70-7 | MFCD00872019 | 99.8% | 376.44 g/mol | C21H28O6 | 1 ML
SDP

Catalog No. 5000227083
Encompass_Preferred

16α-Hydroxyprednisolone is a steroidal metabolite and analytical reference used in drug metabolism and pharmacokinetic studies. It is supplied as a concentrated solution for laboratory use and characterized by its defined molecular identity and high purity.

  • Steroidal metabolite used as a reference standard.
  • Provided as a 10 mM solution in DMSO, 1 mL vial.
  • Purity 99.8% as listed by manufacturer.
  • Molecular formula C21H28O6 and molecular weight 376.44 g/mol.
  • Intended for research use only; not for human or veterinary applications.

Catalog No. 50-002-27083 Supplier Medchemexpress LLC Supplier No. HY11758010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

16α-Hydroxyprednisolone is a steroidal metabolite and analytical reference used in drug metabolism and pharmacokinetic studies. It is supplied as a concentrated solution for laboratory use and characterized by its defined molecular identity and high purity.

  • Steroidal metabolite used as a reference standard.
  • Provided as a 10 mM solution in DMSO, 1 mL vial.
  • Purity 99.8% as listed by manufacturer.
  • Molecular formula C21H28O6 and molecular weight 376.44 g/mol.
  • Intended for research use only; not for human or veterinary applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.