Learn More
Medchemexpress LLC 16α-hydroxyprednisolone | 13951-70-7 | MFCD00872019 | 99.8% | 376.44 g·mol⁻¹ | C21H28O6 | 25 G
SDP
Catalog No. 5000227086
Encompass_Preferred
16α-hydroxyprednisolone is a steroid metabolite used as a reference standard in drug metabolism and pharmacokinetic studies. It is supplied as a solid and is also available as a 10 mM solution in DMSO; the material is provided at high purity for analytical and biochemical applications. Store the solid at 4°C and protect from light.
  • Research-grade steroid metabolite for drug metabolism studies.
  • High purity (≈99.8%) for analytical and biochemical assays.
  • Available as solid and as 10 mM solution in DMSO for assay flexibility.
  • Packaged in a 25 G quantity suitable for larger-scale experimental use.
  • Storage recommendations: solid at 4°C; solutions stored at -80°C (up to 6 months) or -20°C (short term); protect from light.
Catalog No. 50-002-27086 Supplier Medchemexpress LLC Supplier No. HY11758025G
16α-hydroxyprednisolone is a steroid metabolite used as a reference standard in drug metabolism and pharmacokinetic studies. It is supplied as a solid and is also available as a 10 mM solution in DMSO; the material is provided at high purity for analytical and biochemical applications. Store the solid at 4°C and protect from light.
  • Research-grade steroid metabolite for drug metabolism studies.
  • High purity (≈99.8%) for analytical and biochemical assays.
  • Available as solid and as 10 mM solution in DMSO for assay flexibility.
  • Packaged in a 25 G quantity suitable for larger-scale experimental use.
  • Storage recommendations: solid at 4°C; solutions stored at -80°C (up to 6 months) or -20°C (short term); protect from light.