Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 16α-hydroxyprednisolone | 13951-70-7 | MFCD00872019 | 99.8% | 376.44 g/mol | C21H28O6 | 5 G
SDP

Catalog No. 5000226727
Encompass_Preferred

16α-Hydroxyprednisolone is a steroid metabolite supplied as a research reference standard and analytical reagent for metabolic and CYP3A pathway studies. It is provided as a high-purity solid powder intended for analytical and laboratory use.

  • Solid, white to off-white powder.
  • Molecular formula C21H28O6; molecular weight 376.44 g/mol.
  • Purity 99.8% by assay.
  • Packaging available as a 5 G solid package.
  • Storage: solid at 4°C, protect from light.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-26727 Supplier Medchemexpress LLC Supplier No. HY1175805G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

16α-Hydroxyprednisolone is a steroid metabolite supplied as a research reference standard and analytical reagent for metabolic and CYP3A pathway studies. It is provided as a high-purity solid powder intended for analytical and laboratory use.

  • Solid, white to off-white powder.
  • Molecular formula C21H28O6; molecular weight 376.44 g/mol.
  • Purity 99.8% by assay.
  • Packaging available as a 5 G solid package.
  • Storage: solid at 4°C, protect from light.
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.