Learn More
AdipoGen 16-keto-Aspergillimide (1 mg)
SDP
Catalog No. 502053533
Encompass_Preferred
Chemical. CAS: 199784-50-4. Formula: C20H27N3O4. MW: 373.5. Isolated from Aspergillus japonicus Fg-551. Antibiotic. Anthelmintic agent.
Catalog No. 50-205-3533 Supplier AdipoGen Supplier No. AGCN20151M001
Add to Cart
Add to Cart
Chemical. CAS: 199784-50-4. Formula: C20H27N3O4. MW: 373.5. Isolated from Aspergillus japonicus Fg-551. Antibiotic. Anthelmintic agent.