Learn More
Medchemexpress LLC 16-Mercaptopalmitic acid | 69839-68-5 | 288.49 | 250 MG
SDP
Catalog No. 5000351108
Encompass_Preferred
16-Mercaptopalmitic acid is a chain alkylmercaptan that contains a long -CH2 group. It forms a self-assembled monolayer (SAM) on various substrates. It is intended for research use only and is not sold to patients.
  • Chain alkylmercaptan
  • Contains a long -CH2 group
  • Forms a self-assembled monolayer (SAM)
  • Intended for research use only
Catalog No. 50-003-51108 Supplier Medchemexpress LLC Supplier No. 250MGHY137247
16-Mercaptopalmitic acid is a chain alkylmercaptan that contains a long -CH2 group. It forms a self-assembled monolayer (SAM) on various substrates. It is intended for research use only and is not sold to patients.
  • Chain alkylmercaptan
  • Contains a long -CH2 group
  • Forms a self-assembled monolayer (SAM)
  • Intended for research use only