Learn More
Cayman Chemical 16-methyl Stearic Acid, Purity: ≥90%, CAS: 17001-28-4, 5 mg
SDP
Catalog No. 502912824
Encompass_Preferred
A methyl branched-chain fatty acid
Catalog No. 50-291-2824 Supplier Cayman Chemical Supplier No. 364765 MG
A methyl branched-chain fatty acid