Learn More
Cayman Chemical 16-Oxocafestol, Purity: ≥95%, CAS: 108664-98-8, 10 mg
SDP
Catalog No. 502944140
Encompass_Preferred
A derivative of cafestol; increases cytosolic GST activity and acid-soluble sulfhydryl levels in mouse liver and small bowel mucosa at 10 µmol/animal once per day for three days
Catalog No. 50-294-4140 Supplier Cayman Chemical Supplier No. 3994010 MG
A derivative of cafestol; increases cytosolic GST activity and acid-soluble sulfhydryl levels in mouse liver and small bowel mucosa at 10 µmol/animal once per day for three days