Learn More
Cayman Chemical 16-Oxokahweol, Purity: ≥98%, CAS: 108664-99-9, 10 mg
SDP
Catalog No. 502990510
Encompass_Preferred
A synthetic diterpene; increases cytosolic GST activity and acid-soluble sulfhydryl levels in mouse liver and small bowel mucosa at 10 µmol once per day for three days
Catalog No. 50-299-0510 Supplier Cayman Chemical Supplier No. 3011710 MG
A synthetic diterpene; increases cytosolic GST activity and acid-soluble sulfhydryl levels in mouse liver and small bowel mucosa at 10 µmol once per day for three days