Learn More
US Plastic Corporation 16 oz. Polyethylene Funnel - 5" Dia. x 6-1/4"H
SDP
Catalog No. NC1324521
Encompass
These polypropylene and high density polyethylene funnels have many uses. They are inexpensive, lightweight, and durable. They have numerous general applications in the laboratory. The 6-3/8" size has a large throat opening for quick transfer of materials.
Catalog No. NC1324521 Supplier US Plastic Corporation Supplier No. 78023
These polypropylene and high density polyethylene funnels have many uses. They are inexpensive, lightweight, and durable. They have numerous general applications in the laboratory. The 6-3/8" size has a large throat opening for quick transfer of materials.