Learn More
Andwin Scientific 16 OZ PAINT CANS 24/CS
SDP
Catalog No. NC9309721
Encompass_Preferred
The Qorpak tin paint cans have an airtight feature and have an endurance with physical shock. The cans can be sterilized by heat if required. This product has a volume capacity of 16 ounces or 473 mL and is sold in a case of 24.
Catalog No. NC9309721 Supplier Andwin Scientific Supplier No. 1M06Q
The Qorpak tin paint cans have an airtight feature and have an endurance with physical shock. The cans can be sterilized by heat if required. This product has a volume capacity of 16 ounces or 473 mL and is sold in a case of 24.