Learn More
Leica Biosystems 16 OZ SPECIMEN CUPS
Supplier:  Leica Biosystems 3800916
Encompass_Preferred
Specimen Containers are high-density, polyethylene containers that range in size from 4.5 oz to the 192 oz autopsy bucket.Each specimen container has a translucent snap cap lid. These Specimen Containers are ideal for the long term storage of gross tissue.Also available is a 4.5 oz container with screw-on cap.
Catalog No. 50-255-2443