Learn More
Electron Microscopy Sciences 16% Paraformaldehyde (formaldehyde) aqueous solution, 10 bxs/lot
SDP
Catalog No. 50980489
Encompass_Preferred
Paraformaldehyde - methanol free solution. A more efficient and rapid penetrant fixative used in combination with Glutaraldehyde and Acrolein fixatives. Easy to break, prescored, 10 mL ampoules sealed under inert gas 100 mL.
Catalog No. 50-980-489 Supplier Electron Microscopy Sciences Supplier No. 15711
Paraformaldehyde - methanol free solution. A more efficient and rapid penetrant fixative used in combination with Glutaraldehyde and Acrolein fixatives. Easy to break, prescored, 10 mL ampoules sealed under inert gas 100 mL.