Learn More
Electron Microscopy Sciences Beaker Buddy™ 16 Place Holder
SDP
Catalog No. 50949495
Encompass_Preferred
The round, anodized aluminum rack can be used with beakers, water baths, and ice baths. Keep microcentrifuge tubes closed while boiling. Place the tubes in the rack, and then screw on the handle to hold down the cover. The handle is 5 1/8" long, and assist's in the removal from hot beakers. Rack measures 3 1/2" diameter and holds up to 16 tubes.
Catalog No. 50-949-495 Supplier Electron Microscopy Sciences Supplier No. 7236501
The round, anodized aluminum rack can be used with beakers, water baths, and ice baths. Keep microcentrifuge tubes closed while boiling. Place the tubes in the rack, and then screw on the handle to hold down the cover. The handle is 5 1/8" long, and assist's in the removal from hot beakers. Rack measures 3 1/2" diameter and holds up to 16 tubes.