Learn More
Cayman Chemical 16 RHETE Fatty Acids and Deriv
SDP
Catalog No. 502890249
Encompass_Preferred
A minor CYP450 metabolite of arachidonic acid
Catalog No. 50-289-0249 Supplier Cayman Chemical Supplier No. 10004385100 UG
A minor CYP450 metabolite of arachidonic acid