Learn More
Cayman Chemical 16 SHETE Fatty Acids and Deriv
SDP
Catalog No. 502890845
Encompass_Preferred
An inhibitor of proximal tubule ATPase activity; a minor CYP450 metabolite of arachidonic acid
Catalog No. 50-289-0845 Supplier Cayman Chemical Supplier No. 1000438450 UG
An inhibitor of proximal tubule ATPase activity; a minor CYP450 metabolite of arachidonic acid