Learn More
Medique 16 Unit Kit
Catalog No. 17720263 Shop All Fisherbrand Products
Change view
Click to view available options
Material:
Metal
Plastic
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
17-720-263 Plastic
17-720-262 Metal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-720-263
Add to Cart
Add to Cart
ANSI 2021 Class A Unitized kit providing essential first aid supplies for emergencies and everyday safety needs.
  • Includes:
    • 2 - ABD Combine Pads, 5 x 9 in., Sterile
    • 1 - Adhesive Bandages, 3/4 x 3 in. Plastic, 16 ct
    • 2 - Adhesive Bandages, 1 x 3 in. Plastic, 16 ct
    • 1 - Adhesive Bandages, HW Knuckle, 8 ct
    • 1 - Antiseptic BZK Wipes, 10/bx
    • 1 - Burn Cream w/Lidocaine, 0.9 gm pkt, 10/bx
    • 1 - Cold Pack, Small
    • 1 - CPR Mask
    • 1 - Eyewash, Eye Pads and Tape Unit Kit
    • 1 - First Aid Handbook
    • 1 - Gauze Pads 3 x 3 in., Sterile, 4 ct
    • 1 - Gauze Roll 2 in. x 4 yds
    • 1 - Hand Sanitizer, 0.057 oz pkt, 10/bx
    • 1 - Hydrogel Burn Dressing 4 x 4 in.
    • 1 - Nitrile Gloves, Disposable, 2 pr
    • 1 - Rescue Blanket, Silver, 52 x 84 in.
    • 1 - Scissors, 4-1/2 in.
    • 1 - Triangular Bandage w/ Pins, Non-Sterile
    • 1 - Triple Antibiotic Ointment, 0.5 gm pkt, 10/bx
    • 1 - Tweezer, Stainless Steel, 3 in. Slanted Tip
  • Kit fills subject to change
Specifications
Product Type First Aid Kit
Autoclavable Not Autoclavable
Material Plastic
Sterility Non-sterile
Disposable Reusable
Latex Free Yes