Learn More
Ace Glass, Inc. 16 x 150mm tube only. Uses #16 size stopper.
SDP
Catalog No. 50974625
Encompass_Preferred
16x150mm test tube. Test tube uses #16 solid stopper, not included.
Catalog No. 50-974-625 Supplier Ace Glass, Inc. Supplier No. 864506
16x150mm test tube. Test tube uses #16 solid stopper, not included.