Learn More
Kuhner Shaker Inc 16 X ROUNDWELL MTP PORVAIR W
Catalog No. 502570140
Encompass
16 x Roundwell MTP Porvair, white walls, transparent bottom, (24 wells of 3 ml volume, with lid, gamma irradiated)..
Catalog No. 50-257-0140 Supplier Kuhner Shaker Inc Supplier No. SMCR1424F
16 x Roundwell MTP Porvair, white walls, transparent bottom, (24 wells of 3 ml volume, with lid, gamma irradiated)..