Learn More
Sino Biological Recombinant Human RAB27B Protein
Supplier:  Sino Biological 16007HNCH100
Encompass_Preferred
A DNA sequence encoding the human RAB27B (O00194) (Thr2-Lys215) was expressed with two additional amino acids (Gly & Pro) at the N-terminus.
Catalog No. 50-161-9782