Learn More
Sino Biological Recombinant Human KIAA1279 Protein (His & GST Tag)
Supplier:  Sino Biological 16011H20B100
Encompass_Preferred
A DNA sequence encoding the human KIAA1279 (Q96EK5) (Met1-Thr621) was expressed with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-9789