Learn More
Sino Biological Recombinant Human Secretogranin 3 Protein (His Tag)
Supplier:  Sino Biological 16012H08H100
Encompass_Preferred
A DNA sequence encoding the human SCG3 (AAH14539.1) (Met1-Leu468) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-9793