Learn More
Sino Biological Recombinant Human Glycophorin A Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 16018H08H100
Encompass_Preferred
A DNA sequence encoding the human GYPA (P02724) (Met1-Glu91) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-9808