Learn More
Sino Biological Recombinant Human CISD1 Protein (His Tag)
Supplier:  Sino Biological 16024H07E100
Encompass_Preferred
A DNA sequence encoding the human CISD1 (NP_060934.1) (Lys32-Thr108) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9818