Learn More
Sino Biological Recombinant Human Draxin Protein (His Tag)
Supplier:  Sino Biological 16032H08H50
Encompass_Preferred
A DNA sequence encoding the human DRAXIN (NP_940947.3) (Met1-Val349) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9823