Learn More
GFS Chemicals Inc WATERMARK KARL FISCHER TITRANT, STABLE, 5 mg/ml, NON-HYGROSCOPIC | 6 X 1 L | GFS CHEMICALS, INC. Item #: 99667 | Bulk #: 1604
SDP
Catalog No. NC1618102
Encompass_Preferred
WATERMARK KARL FISCHER TITRANT, STABLE, 5 mg/ml, NON-HYGROSCOPIC | For volumetric analysis. Must be used in conjunction with KF Solvent 1609, 1610 or Buffer 1615.
Catalog No. NC1618102 Supplier GFS Chemicals Inc Supplier No. 99667
WATERMARK KARL FISCHER TITRANT, STABLE, 5 mg/ml, NON-HYGROSCOPIC | For volumetric analysis. Must be used in conjunction with KF Solvent 1609, 1610 or Buffer 1615.