Learn More
Sino Biological Recombinant Human FCRL2 Protein (His Tag)
Supplier:  Sino Biological 16041H08H200
Encompass_Preferred
A DNA sequence encoding the human FCRL2 (NP_110391.2) (Met1-Asp395) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9834