Learn More
Sino Biological Recombinant Human Frizzled-10 Protein (hFc Tag)
Supplier:  Sino Biological 16045H02H100
Encompass_Preferred
A DNA sequence encoding the human FZD10 (NP_009128.1) (Met1-Gly161) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9842