Learn More
Sino Biological Recombinant Human ICAM-5 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 16050H08H100
Encompass_Preferred
A DNA sequence encoding the human ICAM5 (NP_003250.3) (Met1-Pro834) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9854