Learn More
Sino Biological Recombinant Human KIR2DL5 Protein (hFc Tag)
Supplier:  Sino Biological 16053H02H200
Encompass_Preferred
A DNA sequence encoding the human KIR2DL5A (NP_065396.1) (Met1-His240) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9856