Learn More
Sino Biological Recombinant Human MEGF10 Protein (hFc Tag)
Supplier:  Sino Biological 16064H02H100
Encompass_Preferred
A DNA sequence encoding the human MEGF10 (XP_016865476.1) (Met1-Gly857) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9867