Learn More
Sino Biological Recombinant Human MEGF10 Protein (His Tag)
Supplier:  Sino Biological 16064H08H100
Encompass_Preferred
A DNA sequence encoding the human MEGF10 (NP_115822.1) (Met1-Gly857) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9869