Learn More
Sino Biological Recombinant Human NCAM2 Protein (hFc Tag)
Supplier:  Sino Biological 16067H02H100
Encompass_Preferred
A DNA sequence encoding the human NCAM2 (NP_004531.2) (Met1-Asn697) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9871