Learn More
Sino Biological Recombinant Human NCAM2 Protein (His Tag)
Supplier:  Sino Biological 16067H08H100
Encompass_Preferred
A DNA sequence encoding the human NCAM2 (NP_004531.2) (Met1-Asn697) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9873