Learn More
Sino Biological Recombinant Human Neuregulin-3/NRG3 Protein (hFc Tag)
Supplier:  Sino Biological 16071H02H100
Encompass_Preferred
A DNA sequence encoding the human NRG3 (NP_001159444.1) (Glu282-Pro342) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9877