Learn More
Sino Biological Recombinant Human RANK/TNFRSF11A Protein (His Tag)
Supplier:  Sino Biological 16078H08H100
Encompass_Preferred
A DNA sequence encoding the human TNFRSF11A (NP_001257879.1) (Met1-Pro212) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9887