Learn More
Sino Biological Recombinant Human BAFFR/TNFRSF13C Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 16079H02H50
Encompass_Preferred
A DNA sequence encoding the human TNFRSF13C (NP_443177.1) (Arg2-Ala71) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9890