Learn More
Sino Biological Recombinant Human FAM20C Protein (His Tag)
Supplier:  Sino Biological 16090H08B100
Encompass_Preferred
A DNA sequence encoding the human FAM20C (NP_064608.2) (Met1-Arg584) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9901