Learn More
Sino Biological Recombinant Human IGFLR1 Protein (hFc Tag)
Supplier:  Sino Biological 16098H02H100
Encompass_Preferred
A DNA sequence encoding the human IGFLR1 (NP_078936.1) (Met1-Pro159) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9903