Learn More
Ace Glass, Inc. 160mL porosity A extraction thimble
SDP
Catalog No. 50973438
Encompass_Preferred
160mL porosity A extraction thimble with an ACE fritted disc sealed in. For use in any standard Soxhlet extraction apparatus.
Catalog No. 50-973-438 Supplier Ace Glass, Inc. Supplier No. 681322
160mL porosity A extraction thimble with an ACE fritted disc sealed in. For use in any standard Soxhlet extraction apparatus.