Learn More
Ace Glass, Inc. 160mL porosity B extraction thimble
SDP
Catalog No. 50973439
Encompass_Preferred
160mL porosity B extraction thimble with an ACE fritted disc sealed in. For use in any standard Soxhlet extraction apparatus.
Catalog No. 50-973-439 Supplier Ace Glass, Inc. Supplier No. 681324
160mL porosity B extraction thimble with an ACE fritted disc sealed in. For use in any standard Soxhlet extraction apparatus.