Learn More
Ace Glass, Inc. 160mL porosity C extraction thimble
SDP
Catalog No. 50978886
Encompass_Preferred
160mL porosity C extraction thimble with an ACE fritted disc sealed in. For use in any standard Soxhlet extraction apparatus.
Catalog No. 50-978-886 Supplier Ace Glass, Inc. Supplier No. 681326
160mL porosity C extraction thimble with an ACE fritted disc sealed in. For use in any standard Soxhlet extraction apparatus.