Learn More
Ace Glass, Inc. 160mL porosity E extraction thimble
SDP
Catalog No. 50973441
Encompass_Preferred
160mL porosity E extraction thimble with an ACE fritted disc sealed in. For use in any standard Soxhlet extraction apparatus.
Catalog No. 50-973-441 Supplier Ace Glass, Inc. Supplier No. 681330
160mL porosity E extraction thimble with an ACE fritted disc sealed in. For use in any standard Soxhlet extraction apparatus.