Learn More
Miele, Inc. ID 160 Injector nozzle. Injector nozzle with plastic supports, Ø 0 1/4 (4), length 160 mm.
Catalog No. NC1302625
Encompass_Preferred
ID 160 Injector nozzle with plastic supports, Ø 0 1/4 (4), length 160 mm.
Suitable for standard mobile injector units and injector baskets
Suitable for measuring flasks and test tubes, for example
Visit www.mieleusa.com/professional for technical information.
Catalog No. NC1302625 Supplier Miele, Inc. Supplier No. 03810350
ID 160 Injector nozzle with plastic supports, Ø 0 1/4 (4), length 160 mm.
Suitable for standard mobile injector units and injector baskets
Suitable for measuring flasks and test tubes, for example
Visit www.mieleusa.com/professional for technical information.