Learn More
Sino Biological Recombinant Human KREMEN1 Protein (His Tag)
Supplier:  Sino Biological 16100H08H50
Encompass_Preferred
A DNA sequence encoding the human KREMEN1 (NP_114434.3) (Met1-Thr394) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9906