Learn More
Sino Biological Recombinant Human RNF43 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 16108H02H100
Encompass_Preferred
A DNA sequence encoding the human RNF43 (NP_060233.3) (Met1-Tyr197) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9907