Learn More
Sino Biological Recombinant Human RNF43 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 16108H08H100
Encompass_Preferred
A DNA sequence encoding the human RNF43 (NP_060233.3) (Met1-Tyr197) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9909