Learn More
Sino Biological Recombinant Human COL6A3 Protein (His Tag)
Supplier:  Sino Biological 16125H07H100
Encompass_Preferred
A DNA sequence encoding the BPTI/Kunitz inhibitor domain of human COL6A3 (P12111-1) (3101T-3177T) was expressed with an N-terminal polyhistidine tag.
Catalog No. 50-161-9921