Learn More
Sino Biological Recombinant Human Frizzled-4 Protein (hFc Tag)
Supplier:  Sino Biological 16137H02H50
Encompass_Preferred
A DNA sequence encoding the human FZD4 (NP_036325.2) (Met1-Glu180) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9930