Learn More
Sino Biological Recombinant Human Frizzled-4 Protein (His Tag)
Supplier:  Sino Biological 16137H08H50
Encompass_Preferred
A DNA sequence encoding the human FZD4 (NP_036325.2) (Met1-Glu180) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9932